Sequential Sterilization Wraps, Halyard
Supplier: O&M HALYARD CANADA, INC CA
10712 10724 10818 10824 10836 10848 68012 68024 68030 68040 68048 68145 68148 68224 68236 68248 68272
CA76540-214CS 102.45 CAD
CA76540-214 CA76540-216 CA76540-218 CA76540-220 CA76540-222 CA76540-224 CA76540-226 CA76540-228 CA76540-230 CA76540-232 CA76540-234 CA76540-236 CA76540-238 CA76540-240 CA76540-242 CA76540-244 CA76540-246 CA76540-248
Sequential Sterilization Wraps, Halyard
Sterilization Wraps
Wrap made of patented low-lint SMS sterilization fabric that is tear-, flame-, lint-, and abrasion-resistant. Recommended for light linen packs, linen gowns, towel packs, single basins, and other light wrapping.
  • Industry standard for sterile processing teams around the world
  • POWERGUARD* technology for enhanced microbial barrier protection
  • Pre-vacuum steam sterilant penetration
  • Ethylene Oxide (EO) sterilant penetration and residuals
  • Class 1 flammability rating
Since 1978, the HALYARD* Sterilization Wrap, made of strong reliable SMS nonwoven fabric, has been helping keep sterilized instruments and supply sets ready for use at a moment’s notice. The company pioneered the development of SMS fabric and today is one of the largest global suppliers. It’s good to know that individual medical and dental practices can also take advantage of the same performance demanded by major medical and surgery centers. All HALYARD* products are made in the USA.
Today, Halyard Health offers six grades of wrap fabric, from H100 to H500, to offer the precise protection required for specific needs. Choosing is easy, the higher the number, the stronger the wrap. HALYARD* is also available in smaller sizes that meet the needs of smaller dental or medical offices.
Certifications: Maintenance of package integrity (MPI) to meet FDA requirements. Instructions for Use to meet FDA guidelines.