Anti-Syntaxin Mouse Monoclonal Antibody [clone: SP8] (Alexa Fluor® 647)

Supplier: Biolegend
827007 827008
76422-678EA 289.14 CAD
76422-678 76422-680
Anti-Syntaxin Mouse Monoclonal Antibody [clone: SP8] (Alexa Fluor® 647)
Antibodies
Order Now

SPECIFICATIONS

Antigen symbol Syntaxin
Environmentally Preferable
Antigen synonyms neuron-specific antigen HPC-1|Syntaxin-1A
Clonality Monoclonal
Conjugation Alexa Fluor® 647
Clone SP8
Reactivity Human,Rat,Mouse
Host Mouse
Isotype IgG1
ImmunoChemistry Yes
Immunogen This monoclonal antibody was raised against a crude synaptic preparation from a post-mortem human brain.
Purification The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
Concentration 0.5 mg/ml
Storage buffer Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide
Storage temperature 2...8 °C
Tested applications IHC-P

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR