Human Recombinant FcεR1alpha (from 293E cells)

Supplier: Biolegend

789102 789104 789106 789108
76422-752EA 162.74 CAD
76422-752 76422-754 76422-756 76422-758
Human Recombinant FcεR1alpha (from 293E cells)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name FcεR1alpha FcεR1alpha
Environmentally Preferable
Protein synonyms Fcε R1alpha|FceR1-alpha|FceR1-a|FceR1a|High affinity immunoglobulin epsilon receptor subunit alpha|FceR1 alpha|high affinity IgE receptor|FCER1A Fcε R1alpha|High affinity IgE receptor|FceR1-alpha|FceR1-a|FceR1a|High affinity immunoglobulin epsilon receptor subunit alpha|FceR1 alpha|FCER1A
Protein/peptide type Recombinant Recombinant
Species Human Human
Source 293E cells 293E cells
Carrier-free Y Y
Concentration 0.2 mg/ml 0.2 mg/ml
Formulation 0.22 µm filtered protein solution is in PBS 0.22 µm filtered protein solution is in PBS
Storage conditions −20…−70 °C −20…−70 °C
Tested applications BA BA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR