AccuThrive Surgical Blades with MicroCoat, Stainless Steel, Sterile, AccuTec Blades
Supplier: Accutec Blades
AVBL-1001-0000 AVBL-1021-0000 AVBL-1031-0000
76539-024BX 582.8 CAD
76539-024 76539-026 76539-028
AccuThrive Surgical Blades with MicroCoat, Stainless Steel, Sterile, AccuTec Blades
Scalpels and Scalpel Assemblies Scalpel Blades
Coated surgical blades for gross dissection.
  • Coated with MicroCoat for sharpness and durability
  • Multiple blade sizes available
AccuThrive's MicroCoat blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp effective edge over multiple uses.