Anti-SARS-CoV-2 Nucleocapsid Human Monoclonal Antibody [clone: HC2003]

Supplier: Genscript
A02039-1 A02039-100
76510-096EA 5052.37 CAD
76510-096 76510-098
Anti-SARS-CoV-2 Nucleocapsid Human Monoclonal Antibody [clone: HC2003]
Antibodies
The product is specific for SARS-CoV-2 Nucleocapsid protein.

  • COVID-19 antibodies
  • Validated for ELISA, WB, FACS and SARS-CoV-2 virus neutralization assay
  • Control antibodies and various antibody pairs available for ELISA-based detection
  • Neutralizing antibody standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples

Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards and control antibodies.

Type: Primary
Antigen: SARS-CoV-2 Nucleocapsid
Clonality: Monoclonal
Clone: HC2003
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR