Clear Seal™ Screw Cap Microtubes
Supplier: MTC BIO, INC.
C3205-C C3205-S C3215-CG C3215-SG C3220-CG C3220-SG C3220-SGL R1081-A
470337-846CS 425 CAD
470337-846 470337-848 470337-850 470337-852 470337-854 470337-856 470337-858 470337-860
Clear Seal™ Screw Cap Microtubes
Tubes Microtubes
Clear Seal™ screw cap microtubes are an all-purpose laboratory microcentrifuge line.
  • Made of crystal clear polypropylene, with leakproof O-ring seal
  • Tubes certified sterile, Rnase-, Dnase-, and pyrogen-free
  • All tubes come conveniently in 20 resealable packs of 50 tubes
ClearSeal™ screw cap microtubes come sterile, in a variety of sizes, cap configurations, graduation options, and even a loop-cap option for the 2.0 ml version. Made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from −90 to +121 °C. All ClearSeal™ screw cap microtubes are sterile, RNase-, DNase-, and pyrogen-free. They may be centrifuged up to 22000×g.