Media/Storage Bottles, Square, Clear PET, United Scientific
Supplier: United Scientific Supplies
BMPE250 BMPE500 BMPE1000
470320-954PK 70.85 CAD
470320-954 470320-956 470320-958
Media/Storage Bottles, Square, Clear PET, United Scientific
Bottles Plastic Bottles
Clear PET bottles for storage.
  • Supplied with polypropylene cap
  • Sterilized using gamma radiation
  • Individually bagged
Clear, square, disposable polyethylene terephthalate (PET) bottles are space-saving and convenient to handle. Useful for mixing, sampling, and storage applications. Includes a blue, liner-less polypropylene plug-seal autoclavable cap and features raised graduations. Neck int.Ø is 37 mm.
PET is transparent, lightweight, and break-resistant. Bottles can be used in environments ranging from −40...+50 °C. PET cannot be autoclaved.