Media/Storage Bottles, United Scientific
Supplier: United Scientific Supplies
BM0100 BM0250 BM0500 BM1000 BM2000 BM5000 BM10000 BM20000
470332-664PK 182.8 CAD
470332-664 470332-666 470332-668 470332-670 470332-672 470332-674 470332-676 470332-678
Media/Storage Bottles, United Scientific
Bottles Glass Bottles
General-purpose media bottles.
  • Autoclavable
  • Borosilicate glass bottles
  • Supplied with polypropylene cap
These borosilicate glass general-purpose media bottles are autoclavable with permanent white imprinted graduations and marking spots. The screw thread opening has an int.Ø of approximately 30 mm. The supplied GL 45 cap is linerless, and both the cap and drip-free ring are polypropylene and autoclavable to 140 °C. Maximum use temperature of the bottles is 450 °C.