BRAND® Tip Boxes for BRAND® Bulk Tips and Tip Refills, BrandTech
Supplier: Brandtech
732990 732994 732996
76534-684EA 13.56 CAD
76534-684 76534-686 76534-688 76534-690
BRAND® Tip Boxes for BRAND® Bulk Tips and Tip Refills, BrandTech
Pipette Tip Boxes and Racks
Empty polypropylene tip boxes with carrier plates to fit BRAND® pipette tips.
  • Refillable
  • Autoclavable
  • Color-coded for easy identification
BRAND® Tip Boxes are designed to hold BRAND® bulk tips and Tip Refills. Carrier plate is included for bulk tips. Gray carrier plate is designed to fit nano-cap™, ultra-micro, and 50 µl tips, in both unfiltered and filtered variations. Yellow carrier plate is designed to fit BRAND 200 µl tips. Green carrier plate is for use with 300 µl tips. The box with blue carrier plate has extra height to fit 1000 µl tips, but will accept carrier plates from other tip sizes. Box and carrier plate are fully autoclavable at 121 °C for 20 minutes for sterilization.
Ordering information: Box/carrier plate is sold individually.
Packaging: Recyclable packaging.