Wind Up Tape Measure
Supplier: EISCO SCIENTIFIC LLC
PH0076F
470337-816EA 48 CAD
470337-816
Wind Up Tape Measure
Measuring Tapes
This flexible tape measure comes conveniently in a wind-up roll for easy use.
  • Made of non-conducting material
  • Waterproof, washable, and water-resistant
  • Length of 30 meters
Tape is made of fiberglass with PVC covering. One side is in metric and the other in feet/inches. The case is made of a zinc-coated steel sheet, covered with vinyl and fitted with a flush winding-handle.