VWR® Mega Star 600 and 600R Small Bench Centrifuges and Packages
Supplier: VWR International
76533-880 76533-882 76533-884 76533-886
76533-880EA 4376.6 CAD
76533-880 76533-882 76533-884 76533-886
VWR® Mega Star 600 and 600R Small Bench Centrifuges and Packages
Centrifuges Benchtop Centrifuges
VWR® Mega Star 600 Ventilated and 600R Refrigerated Small Bench Centrifuge optimize bench space with maximized capacity in a compact footprint. Available in both a ventilated and refrigerated option, units come with a user-friendly interface and are adaptable in the ever evolving clinical and research laboratory.
  • Auto-Lock™ Rotor Exchange System: Units have secure, push-button Auto-Lock™ rotor exchange
  • Swap out rotors in as little as three seconds for trouble-free installation/removal and clear chamber access for cleaning convenience. No tools required
  • ClickSeal™ Biocontainment solution: The centrifuge packages include biocontainment sealing options, including certified ClickSeal™ lids for glove-friendly, one-handed operation
  • Biocontainment certification by the Public Health England, Porton Down, UK
  • Space-saving design, the compact dimensions optimize work area
  • Ergonomic height and curved design ensure easy sample loading/unloading and easy cleaning
Certifications: ANSI/UL 61010-1
UL 61010-2-020
Ordering information: Both unit only and package options are available. If centrifuge only option is purchased, accessories must be purchased separately. Packages include centrifuge plus TX-150 swinging bucket rotor, 4× buckets, 4× ClickSeal biocontainment lids.