LunaScript Multiplex One-Step RT-PCR Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E1555S E1555L
CA76522-054EA 377 CAD
CA76522-054 CA76522-056
LunaScript Multiplex One-Step RT-PCR Kit, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The LunaScript Multiplex One-Step RT-PCR Kit offers a streamlined protocol for cDNA synthesis and PCR amplification in a single reaction.
  • Simplified workflow with cDNA synthesis and PCR amplification in a single, closed-tube reaction
  • Highly sensitive - detect as low as 0.01 pg of human total RNA
  • Superior multiplexing capacity
  • Dual control of enzyme activity via reversible, aptamer-based inhibition for room temperature setup
Kit contains a reaction mix and an enzyme mix featuring Luna WarmStart Reverse Transcriptase and Q5 Hot Start High Fidelity DNA polymerase. The kit is optimized for multiplex target amplification to enable diagnostics, pathogen detection, and viral genome sequencing.