NEBNext Ultra II DNA PCR-free Library Prep Kit for Illumina, New England Biolabs
Supplier: New England Biolabs (NEB)
E7410S E7410L
CA76538-656EA 876 CAD
CA76538-656 CA76538-658
NEBNext Ultra II DNA PCR-free Library Prep Kit for Illumina, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Ultra II DNA PCR-free Library Prep Kit contains the enzymes and buffers required to convert a broad range of pre-fragmented DNA input amounts into libraries for next-generation sequencing on the Illumina platform, all without an amplification step.
  • Save time with an optimized PCR-free workflow, reduced hands-on time and automation compatibility
  • Generate high quality libraries without an amplification step with inputs from 250 to 1000 ng
  • Improved library uniformityFor use with NEBNext Unique Dual Index UMI Adaptors DNA Set 1
  • Also compatible with other Illumina-compatible index adaptors with 5’ single T overhang
The fast, user-friendly workflow requires minimal hands-on time, and maintains excellent library yields and GC coverage without requiring PCR. Don’t be biased! The NEBNext® Ultra™ II DNA PCR-free Library Prep Kit for Illumina® can save you time and minimize amplification-induced sequence bias and errors with a complete solution for PCR-free NGS library preparation. Built on the robust and reliable Ultra II reagents and workflow from New England Biolabs®, you can trust that your samples are in the best hands – your own.