NEBridge Ligase Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
M1100S M1100L
CA76538-670EA 142 CAD
CA76538-670 CA77025-080
NEBridge Ligase Master Mix, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
NEBridge Ligase Master Mix is a 3X master mix for Golden Gate Assembly. Designed for use with NEB Type IIS restriction enzymes, this master mix contains T4 DNA Ligase in an optimized reaction buffer with a proprietary ligation enhancer.
  • Optimized for efficient and accurate Golden Gate Assembly
  • Convenient 3X Master Mix formatUse with NEB Type IIS Restriction Enzymes
  • Use for seamless cloning –no scar remains following assembly
  • Ideal for ordered assembly of multiple fragments (2 to 50+) in a single reaction
Users need only choose their preferred NEB Type IIS restriction enzyme and add DNA substrates to be assembled. Low complexity single fragment insertions, as well as moderate complexity (3 to 6 fragment) and high complexity (7 to 25 fragment) assemblies, are all supported with this optimized reagent and accompanying protocols.