CELLvo™ Osteoarthritic Human Chondrocytes P0 (50-70y), StemBioSys
Supplier: StemBioSys
OA-100M-000
OA-100M-6WP
OA-100M-T75
OA-100M-T150
OA-100F-000
OA-100F-6WP
OA-100F-T75
OA-100F-T150
76475-914EA
2869.52
CAD
76475-914
76475-916
76475-918
76475-920
76475-922
76475-924
76475-926
76475-928
CELLvo™ Osteoarthritic Human Chondrocytes P0 (50-70y), StemBioSys
Cells
CELLvo™ Ostoeoarthritic Human Chondrocytes (50-70y) are primary, uncultured cells isolated from recently-diseased cadaver cartilage from donors between 50 and 70 years old. Osteoarthritis status is determined by macroscopic inspection using a modified Outerbridge (1960) scale. Cells are isolated by enzymatic digestion prior to cryopreservation.