CELLvo™ Healthy Human Chondrocytes P0 (50-70y), StemBioSys
Supplier: StemBioSys
HCA3-100M-000
HCA3-100M-6WP
HCA3-100M-T75
HCA3-100M-T150
HCA3-100F-000
HCA3-100F-6WP
HCA3-100F-T75
HCA3-100F-T150
76475-898EA
1913.02
CAD
76475-898
76475-900
76475-902
76475-904
76475-906
76475-908
76475-910
76475-912
CELLvo™ Healthy Human Chondrocytes P0 (50-70y), StemBioSys
Cells
CELLvo™ Healthy Human Chondrocytes (50-70y) are primary, uncultured cells isolated from recently-diseased cadaver cartilage from donors between 50 and 70 years old. Healthy status is determined by macroscopic inspection of the tissue using a modified outerbridge (1960) scale in order to distinguish healthy joints from those with signs of osteoarthritis. Cells are isolated by enzymatic digestion prior to cryopreservation.