arium® RO Pre-treatment Cartridge, Sartorius
Supplier: Sartorius
arium®
H2O-CPFAD-1 H2O-CPFAD-1
CA76533-170PK 653.36 CAD
76533-170 CA76533-170
arium® RO Pre-treatment Cartridge, Sartorius
Water Purification Systems Pure and Ultra Pure Water Systems
Reliably removes oxidizing agents, heavy metals, and particular impurities.
  • Fast and effective adsorption of impurities through high-grade activated carbon
  • 5 μm depth filter for the retention of particles
  • Highly efficient catalyst for removing free chlorine
  • Patented cartridge design for easy installation
An efficient protection for a downstream reverse osmosis (RO) membrane is the combination of spherical, catalytic effective activated carbon, a catalyst, and a depth filter.The pre-treatment cartridge reliably removes oxidation agents, such as free chlorine, heavy metal ions, and particulate contaminants from the feed water of the system.
This cartridge is designed specifically for use with arium® Advance RO, arium® Advance EDI, and arium® 61316 and 61215 systems.
Materials of construction: High-quality polypropylene