Test Tubes, Polypropylene, United Scientific
Supplier: United Scientific Supplies
TPP1275 TPP1275-PK/250
470332-800PK 14.15 CAD
470332-800 470332-802
Test Tubes, Polypropylene, United Scientific
Tubes Test Tubes
Round-bottom polypropylene test tubes.
  • 12×75 mm tubes
  • 5 ml capacity
  • Autoclavable
Tubes are available in packs of 100 or 250.