Dynamics Carts, United Scientific
Supplier: United Scientific Supplies
DNCART2
470332-618EA 29.5 CAD
470332-618
Dynamics Carts, United Scientific
Educational Materials Physics Educational Materials Mechanics Learning Activities
Set of two dynamics carts to demonstrate energy conservation, velocity, elastic collisions, acceleration, and momentum.
  • Designed with durable spring steel bumpers and rubber stoppers
  • Features convenient wells to accommodate additional masses
  • Activity guide included
Ideal for demonstrating experiments in conservation, velocity, elastic collisions, acceleration, and momentum. Included are two impact carts and two spring steel bumpers with rubber stopper assemblies. Each cart features a well for additional masses (not included).