Unitd Scientific™ Van De Graaff Generator, Small Electric
Supplier: United Scientific Supplies
VANDGF1-SE VANDGF1-SEB
470332-628EA 537.25 CAD
470332-628 470332-630
Unitd Scientific™ Van De Graaff Generator, Small Electric
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This excellent quality electrostatic generator consists of a charge generation system, an aluminum collector sphere, and an electric motor drive on a sturdy base.
  • Requires a 110 VAC outlet
  • Includes activity guide
The charge generation system is based on a moving neoprene belt mounted on insulating rollers with metal charge transfer combs. The system is housed in a clear tube that allows for easy viewing of the moving parts. A 100,000 volt potential can be generated.