Salus HC PAPR Kits, Bullard
Supplier: Bullard
SALHC20LFLHE SALHC20LFLHE2 SALHC20LF2M SALHC20LF2L SALHC20LF2L2 SALHC20LFMHE2 SALHC20LFMHE SALHC20LF2M2
76534-568EA 3239.32 CAD
76534-568 76534-566 76534-574 76534-564 76534-572 76534-562 76534-570 76534-560
Salus HC PAPR Kits, Bullard
Respirators Respiratory Systems
Ideal for healthcare and pharmaceutical applications.
  • Unique shoulder carriage design for maximum comfort
  • Auto dampening technology to minimize noise
  • Closed cell foam; easy to clean
Unit features one-button operation located on chest, dual alarms, and two-speed blower. Comfortable hood cosntructed of Tychem 2000. Small, lightweight battery can easily be changed while operating the blower unit.
Certifications: NIOSH approved to P100 filter standard.