Volumetric Flasks, Class A, with Glass Stopper, Batch Certified, QR, United Scientific Supplies
Supplier: United Scientific Supplies
FG5640-1000QR FG5640-100QR FG5640-10QR FG5640-2000QR FG5640-200QR FG5640-20QR FG5640-250QR FG5640-25QR FG5640-500QR FG5640-50QR FG5640-5QR
76531-868EA 85.73 CAD
76531-868 76531-870 76531-872 76531-880 76531-882 76531-884 76531-890 76531-892 76531-900 76531-902 76531-904
Volumetric Flasks, Class A, with Glass Stopper, Batch Certified, QR, United Scientific Supplies
Flasks Volumetric Flasks
Durable, machine-blown body with heavy beaded tubing neck.
  • Sharp graduation line and large white block lettering
A printed QR code allows for easy access to the calibration certificate from a mobile device or computer. A paper certificate is also included.
Certifications: Complies with ASTM E288 Class A specifications; batch certified.
Ordering information: A ground glass stopper is included.