Volumetric Flasks, Class B, with Snap Cap, United Scientific Supplies
Supplier: United Scientific Supplies
FG5581-100 FG5581-1000 FG5581-2000 FG5581-250 FG5581-50 FG5581-500
76531-806PK 233.39 CAD
76531-806 76531-808 76531-810 76531-812 76531-814 76531-816
Volumetric Flasks, Class B, with Snap Cap, United Scientific Supplies
Flasks Volumetric Flasks
Durable, machine-blown body with heavy beaded tubing neck.
  • Sharp graduation line and large white block lettering
  • Flasks meet Class B specifications
Plastic snap caps are included.