Volumetric Flasks, Class A, with Snap Cap, Batch Certified, QR, United Scientific Supplies
Supplier: United Scientific Supplies
FG5580-1000QR FG5580-100QR FG5580-2000QR FG5580-200QR FG5580-250QR FG5580-500QR FG5580-50QR
76531-782EA 69.42 CAD
76531-782 76531-784 76531-790 76531-792 76531-796 76531-802 76531-804
Volumetric Flasks, Class A, with Snap Cap, Batch Certified, QR, United Scientific Supplies
Flasks Volumetric Flasks
Durable, machine-blown body with heavy beaded tubing neck.
  • Sharp graduation line and large white block lettering
A printed QR code allows for easy access to the calibration certificate from a mobile device or computer. A paper certificate is also included.
Certifications: Complies with ASTM E288 Class A specifications; batch certified.
Ordering information: A plastic snap cap is included.