Volumetric Flask, Class A, Snap Cap, United Scientific Supplies
Supplier: United Scientific Supplies
FG5580-100 FG5580-1000 FG5580-200 FG5580-2000 FG5580-250 FG5580-50 FG5580-500
76531-778EA 28.45 CAD
76531-778 76531-780 76531-786 76531-788 76531-794 76531-798 76531-800
Volumetric Flask, Class A, Snap Cap, United Scientific Supplies
Flasks Volumetric Flasks
Durable, machine-blown body with heavy beaded tubing neck.
  • Sharp graduation line and large white block lettering
A plastic snap cap is included.
Certifications: Complies with ASTM E288 Class A specifications; batch certified.