Erlenmeyer Flasks, with Screw Cap, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
FG5021-100 FG5021-1000 FG5021-150 FG5021-250 FG5021-500
76531-746PK 96.85 CAD
76531-746 76531-748 76531-750 76531-752 76531-754
Erlenmeyer Flasks, with Screw Cap, Borosilicate Glass, United Scientific Supplies
Flasks Erlenmeyer Flasks
Erlenmeyer flasks with autoclavable blue screw caps with liner.
  • Made from borosilicate glass for an excellent combination of thermal shock resistance and physical strength
  • Feature heavy-duty rims and durable white enamel graduations
Recommended for culturing, mixing and storing applications. Borosilicate glass flasks are designed to give the best combination of thermal shock resistance and physical strength.