Boiling Flasks, Round Bottom, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
FG4260-100 FG4260-1000 FG4260-125 FG4260-150 FG4260-2000 FG4260-25 FG4260-250 FG4260-300 FG4260-3000 FG4260-50 FG4260-500
76531-694PK 41.66 CAD
76531-694 76531-696 76531-698 76531-700 76531-702 76531-704 76531-706 76531-708 76531-710 76531-712 76531-714
Boiling Flasks, Round Bottom, Borosilicate Glass, United Scientific Supplies
Flasks Round Bottom Flasks
Boiling flasks designed for heating mantles.
  • Uniform wall thickness
  • Low expansion borosilicate glass
Made from low-expansion borosilicate glass, these flasks feature uniform wall thickness for superior mechanical strength and shock resistance.