Boiling Flasks, Flat Bottom, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
FG4060-100 FG4060-1000 FG4060-150 FG4060-2000 FG4060-250 FG4060-300 FG4060-3000 FG4060-50 FG4060-500 FG4060-5000
76531-674PK 41.66 CAD
76531-674 76531-676 76531-678 76531-680 76531-682 76531-684 76531-686 76531-688 76531-690 76531-692
Boiling Flasks, Flat Bottom, Borosilicate Glass, United Scientific Supplies
Flasks Flat Bottom Flasks
Flat bottom flasks for resting on the tabletop.
  • Uniform wall thickness
  • Low expansion borosilicate glass
These boiling flasks are made from low-expansion borosilicate glass. Feature uniform wall thickness for superior mechanical strength and shock resistance.