Disposable Culture Tubes, Plain, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
DCT051-0650 DCT051-1075 DCT051-1275 DCT051-1300 DCT051-1585 DCT051-1610 DCT051-1625 DCT051-1650 DCT051-1850 DCT051-20150 DCT051-25150
76531-630CS 198.38 CAD
76531-630 76531-632 76531-634 76531-636 76531-638 76531-640 76531-642 76531-644 76531-646 76531-648 76531-650
Disposable Culture Tubes, Plain, Borosilicate Glass, United Scientific Supplies
Tubes Culture Tubes
Disposable culture tubes manufactured from high quality 5.1 expansion borosilicate glass.
  • Rimless design for easy plugging with stopper (not included)
  • Packaged in shrinkwrapped boxes
Caps not included.
Certifications: Meet USP Type I and ASTM E438, Type I, Class B requirements.
Packaging: Tubes are packaged in shrink-wrapped boxes to keep them particle-free, and four boxes are included in each case.