Culture Tubes with Cap, Round Bottom, United Scientific Supplies
Supplier: United Scientific Supplies
TT9900-10 TT9900-30 TT9900-5 TT9900-60
76532-238PK 23.97 CAD
76532-238 76532-240 76532-242 76532-244
Culture Tubes with Cap, Round Bottom, United Scientific Supplies
Tubes Culture Tubes
Round-bottom culture tubes manufactured from ASTM E438 Type 1 Class A borosilicate 3.3 glass.
  • Autoclavable
Include polypropylene screw caps with PTFE liner.