Separatory Funnels, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
SF149-100 SF149-1000 SF149-125 SF149-2000 SF149-250 SF149-50 SF149-500
76532-178PK 103.11 CAD
76532-178 76532-180 76532-182 76532-184 76532-186 76532-188 76532-190
Separatory Funnels, Borosilicate Glass, United Scientific Supplies
Funnels Separating Funnels
These pear-shaped borosilicate glass funnels include a PTFE stopcock and a plastic stopper.
  • Feature a marking spot and large white block letters for easy identification of capacity, as per ASTM E1096
PTFE stopcocks are compliant with ASTM standard E675.