Pipettes, Transfer, Volumetric, Class B, United Scientific Supplies
Supplier: United Scientific Supplies
PT7102-A PT7102-B PT7102-C PT7102-D PT7102-E PT7102-F PT7102-G PT7102-H
76532-162PK 38.46 CAD
76532-162 76532-164 76532-166 76532-168 76532-170 76532-172 76532-174 76532-176
Pipettes, Transfer, Volumetric, Class B, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
Volumetric pipettes with color-coding.
  • Printed with durable amber markings
  • Color-coded in blue for easy identification of capacity
Made from borosilicate glass in compliance with ASTM E969, Class B standards; calibrated to deliver.