Pipettes, Transfer, Volumetric, Class A, Individually Certified, United Scientific Supplies
Supplier: United Scientific Supplies
PT7100-A PT7100-B PT7100-C PT7100-D PT7100-E PT7100-F PT7100-G PT7100-H
76532-130PK 140.96 CAD
76532-130 76532-132 76532-134 76532-136 76532-138 76532-140 76532-142 76532-144
Pipettes, Transfer, Volumetric, Class A, Individually Certified, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
Volumetric pipettes with color-coding.
  • Printed with durable amber markings
  • Color-coded in blue for easy identification of capacity
Made from borosilicate glass in compliance with ASTM E969, Class A standards. Individually serialized and certified; calibrated to deliver.