Boiling Flasks, Round Bottom, Ground Glass Joints, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
FRB057-100 FRB057-1000 FRB057-2000 FRB057-250 FRB057-3000 FRB057-50 FRB057-500 FRB057-5000
76531-996PK 87.29 CAD
76531-996 76531-998 76532-000 76532-002 76532-004 76532-006 76532-008 76532-010
Boiling Flasks, Round Bottom, Ground Glass Joints, Borosilicate Glass, United Scientific Supplies
Flasks Round Bottom Flasks
Round-bottom boiling flasks with ground glass joints (24/40).
  • Made from low-expansion borosilicate glass
Feature uniform wall thickness for superior mechanical strength and shock resistance.
Order Now
SPECIFICATIONS
Environmentally Preferable