Filtration, All-Glass Vacuum Filter Holder Set, 90 mm, United Scientific Supplies
Supplier: United Scientific Supplies
FHFG1000-VWR FHBT2000 FHBT5000 FHGBC90 FHGF1000 FHGSP90 FHSC90 FHSS90 FHSSR90
76293-574EA 170.68 CAD
76293-574 76531-956 76531-958 76531-966 76531-970 76531-974 76531-984 76531-988 76531-994
Filtration, All-Glass Vacuum Filter Holder Set, 90 mm, United Scientific Supplies
Filter Holders
This all-glass filter holder is used in particulate contamination analysis for filtering aqueous, organic, or corrosive liquids. It is also recommended for HPLC solvent filtration.
  • Borosilicate glass funnel
  • PTFEcoated stainless steel screen
Complete set includes a borosilicate glass funnel, base and tubulated cap, anodized aluminum spring clamp, and PTFEcoated stainless steel screen. Membrane filters are not included. Parts are also available separately.
Specifications
Materials: Borosilicate glass funnel, base and tubulated cap; anodized aluminumMore Product Information