Graduated Cylinders, Glass, Class A, Individually Certified, with Stopper, United Scientific Supplies
Supplier: United Scientific Supplies
CY2980-10 CY2980-100 CY2980-1000 CY2980-2000 CY2980-25 CY2980-250 CY2980-50 CY2980-500
76531-554EA 65.8 CAD
76531-554 76531-556 76531-558 76531-560 76531-562 76531-564 76531-566 76531-568
Graduated Cylinders, Glass, Class A, Individually Certified, with Stopper, United Scientific Supplies
Cylinders Measuring Cylinders
Borosilicate glass cylinders feature heavy uniform wall tubing and strong, stable, hexagonal bases. All cylinders come with bumper guards.
  • With hollow glass stopper
  • Double metric scale, calibrated to contain
Cylinders comply with ASTM E1272, Class A, meets USP standards, individually serialized and certified.