Condenser, Liebig, United Scientific Supplies
Supplier: United Scientific Supplies
CNL200 CNL300 CNL400 CNL500
76531-544EA 66.43 CAD
76531-544 76531-546 76531-548 76531-550
Condenser, Liebig, United Scientific Supplies
Condensers
Liebig condenser, used to cool and condense hot vapor as part of a distilling apparatus.
  • 24/40 socket with screw thread connector
This condenser has a straight inner tube that the condensing vapor passes through, and a larger outer jacket that cooling water passes through.