Crucibles, Glass, United Scientific Supplies
Supplier: United Scientific Supplies
CGQ015 CGQ015-L CGQ025 CGQ025-L CGQ050 CGQ050-L CGQ080 CGQ080-L CGQ100 CGQ100-L CGQ150 CGQ150-L
76531-520EA 63.38 CAD
76531-520 76531-522 76531-524 76531-526 76531-528 76531-530 76531-532 76531-534 76531-536 76531-538 76531-540 76531-542
Crucibles, Glass, United Scientific Supplies
Crucibles
Crafted from clear quartz glass, offering low expansion, dimensional stability and thermal shock resistance.
  • Maximum operating temperature: 1050 °C (1922 °F)
  • Quartz lid sold separately
Ideal for analytical applications that require ashing of samples.