Media/Storage Bottles, GL 80, Wide Mouth, Square, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
BSWM1000 BSWM2000 BSWM500 BSWM5000
76531-508PK 250.47 CAD
76531-508 76531-510 76531-512 76531-514
Media/Storage Bottles, GL 80, Wide Mouth, Square, Borosilicate Glass, United Scientific Supplies
Bottles Glass Bottles
Wide mouth, square-shaped, media bottles have large openings that enable the easy storage and transfer of substances.
  • Feature printed white graduations (from 200 to 1000 ml, at 100 ml intervals) and durable marking spots
  • GL 80 polypropylene screw caps included
  • Autoclavable
Every bottle is autoclavable and comes with easy to read imprinted graduations.