Automatic Burette, Class A, 10 ml, Individually Certified, United Scientific Supplies
Supplier: United Scientific Supplies
BRA217-10 BRA217-25 BRA217-50 BRB0500 BRB2000
76531-498EA 378.29 CAD
76531-498 76531-500 76531-502 76531-504 76531-506
Automatic Burette, Class A, 10 ml, Individually Certified, United Scientific Supplies
Burettes
Self-zeroing burettes designed to mount on glass bottle with 24/29 joints (sold separately).
  • Manufactured from precision bore tubing
  • Features amber graduations and Schellbach backing for easy reading of liquid levels
Class A, per ASTM E-287, meets USP standards. Individually serialized and certified. Manufactured from precision bore tubing and calibrated to deliver (TD, Ex) on computer-controlled machines.
Includes a PTFE needle valve stopcock, intermediate stopcock and rubber bellows.