Burette, Class A, PTFE Stopcock, Batch Certified, United Scientific Supplies
Supplier: United Scientific Supplies
BR2129-10 BR2129-25 BR2129-50
76531-484EA 92.81 CAD
76531-484 76531-486 76531-488
Burette, Class A, PTFE Stopcock, Batch Certified, United Scientific Supplies
Burettes
Class A burette, per ASTM E287. Meets USP standards and is batch certified.
  • Graduated at intervals with a waiting time of 30 seconds
  • Includes PTFE straight bore stopcock
  • Available in multiple sizes
Manufactured from precision bore tubing, calibrated to deliver (TD, Ex) on computer-controlled machines.