Burette, Class A, PTFE Stopcock, Individually Certified, United Scientific Supplies
Supplier: United Scientific Supplies
BR2128-10 BR2128-25 BR2128-50
76531-478EA 184.52 CAD
76531-478 76531-480 76531-482
Burette, Class A, PTFE Stopcock, Individually Certified, United Scientific Supplies
Burettes
Class A burette, per ASTM E287. Meets USP standards and is individually certified.
  • Graduated at intervals with a waiting time of 30 seconds
  • Includes PTFE straight bore stopcock
  • Available in multiple sizes
Manufactured from precision bore tubing, calibrated to deliver (TD, Ex) on computer-controlled machines.