Burette, Class A, Screw Thread Stopcock, Batch Certified, United Scientific Supplies
Supplier: United Scientific Supplies
BR2121-10 BR2121-25 BR2121-50
76531-462EA 67.58 CAD
76531-462 76531-464 76531-466
Burette, Class A, Screw Thread Stopcock, Batch Certified, United Scientific Supplies
Burettes
Class A burette, per ASTM E287. Meets USP standards and is batch certified.
  • Graduated at intervals with a waiting time of 30 seconds
  • Includes PTFE screw thread stopcock
  • Available in multiple sizes
Manufactured from precision bore tubing, calibrated to deliver (TD, Ex) on computer-controlled machines.