Burettes, Class B, with PTFE Detachable Stopcocks, United Scientific Supplies
Supplier: United Scientific Supplies
BR2116-10 BR2116-25 BR2116-50
76531-450EA 99.89 CAD
76531-450 76531-452 76531-454
Burettes, Class B, with PTFE Detachable Stopcocks, United Scientific Supplies
Burettes
Made from high-quality accurate bore tubing for a high degree of accuracy, reliability and durability.
  • Class B burette
  • Graduated at intervals
  • Available in multiple sizes
Detachable PTFE stopcock and tip facilitate easy cleaning and convenient replacement of broken tips.