Media/Storage Bottles, Square, United Scientific Supplies
Supplier: United Scientific Supplies
BM0100-S BMS0250 BMS0500 BMS1000
76531-424EA 2.97 CAD
76531-424 76531-434 76531-436 76531-438
Media/Storage Bottles, Square, United Scientific Supplies
Bottles Glass Bottles
Pack of 10 square-shaped, autoclavable borosilicate glass media / storage bottles.
  • Feature printed white graduations (from 50 - 200 ml) and durable marking spots
  • Screw thread opening has a 30 mm inner diameter
  • Maximum use temperature is 450 °C (842 °F)
  • Autoclavable
Included GL 45 cap is linerles, both the caps and drip-free rings are made from polypropylene and autoclavable to 140 °C.