Beakers, Low Form, Heavy Duty, United Scientific Supplies
Supplier: United Scientific Supplies
BG1003-100 BG1003-1000 BG1003-150 BG1003-2000 BG1003-250 BG1003-400 BG1003-4000 BG1003-600 BG1003-50
76531-378PK 107.61 CAD
76531-378 76531-380 76531-382 76531-384 76531-386 76531-388 76531-390 76531-392 470349-762
Beakers, Low Form, Heavy Duty, United Scientific Supplies
Beakers
Made from low-expansion borosilicate glass with extra-thick walls, these beakers are ideal for tough handling conditions, including mechanized washing.
  • Include white printed graduations (from 20 - 80 ml, at 10 ml intervals) and durable marking area
  • Tolerance is approximately +/- 5%
Available in multiple sizes.