Buchner Funnel With Fritted Disc, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
BF256-15C BF256-15F BF256-15M BF256-30C BF256-30F BF256-30M BF256-60C BF256-60F BF256-60M
76531-324PK 47.05 CAD
76531-324 76531-326 76531-328 76531-330 76531-332 76531-334 76531-336 76531-338 76531-340
Buchner Funnel With Fritted Disc, Borosilicate Glass, United Scientific Supplies
Funnels Filtration Funnels Büchner Funnels
Pack of 2 thick-walled borosilicate glass Buchner funnels.
  • Include fritted discs
  • Funnels feature beaded rims
Porosity of course (C) is 40 - 60 microns, porosity of fine (F) is 5 - 15 microns, porosity of medium (M) is 15 - 40 microns.