Orum International TRIO.BAS™ FLO.GAS Gas Testing System
Supplier: HARDY DIAGNOSTICS CA
BAS597 BAS598
CA76509-310EA 16076.2 CAD
CA76509-310 CA76509-312
Orum International TRIO.BAS™ FLO.GAS Gas Testing System
Samplers Gas Samplers
The FLO.GAS system tests for the presence of microorganisms in compressed air and nitrogen/gas samples supplied from tanks and pipes under pressure.
  • Available in Petri plate or contact plate format
  • Can be used independently or with a TRIO.BAS™ 100 L/min air sampler
Included is the instrument with calibrated flow meter, stainless steel AISI 316 bell chamber, ASPIGAS chamber, stainless steel aspirating head for Petri plates, and digital timer.