Science Take-Out® Nicotine Addiction: Breaking Up with My Vape
Supplier: SCIENCE TAKE-OUT, LLC
STO-167-NOTG STO-167U STO-167
470337-446KT 25.65 CAD
470337-446 470337-447 470337-448
Science Take-Out® Nicotine Addiction: Breaking Up with My Vape
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Why is it so difficult to quit vaping?
  • Available as a single kit with materials for 1 student or group
  • Unassembled 10 pack for additional savings
  • No additional equipment needed
Follow the story of a teen who is trying to quit vaping. Model how vaping nicotine affects the receptors and dopamine levels in the brain reward pathway. Use the model to understand nicotine tolerance, dependence, addiction, withdrawal and recovery from nicotine addiction.