dNTP Mix, Biotium
Supplier: Biotium
40053 40054
10001-640EA 446.05 CAD
10001-640 10001-642
dNTP Mix, Biotium
Nucleic Acid Reagents Nucleotides
Highly purified mix of dNTPs (>99% by HPLC) in 0.6 mM Tris-HCl (pH 7.5); each dNTP is at a concentration of 10 or 25 mm in the solution provided.
The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.